Adrenalone hydrochloride, hydrat ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

A1234-25G A1234-5G
TCIAA1234-25GEA 115 EUR
TCIAA1234-25G TCIAA1234-5G
Adrenalone hydrochloride, hydrat ≥98.0% (by HPLC, titration analysis)
Adrenalone hydrochloride
Formule: C₉H₁₁NO₃·HCl·xH₂O
MDL Nummer: MFCD00035075
CAS nummer: 62-13-5
EINECS: 200-525-7
Merck Index: 14,00171
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40b86b009e54f15a1a5082d6bfe878d5210b0baf&vid=664092f2d9fcaf12c7ce17cdb7c1d9fd81d38814&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0% (calcd.on anh.substance)
Solubility in Water almost transparency
Water max. 8.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR