N-(4-Methoxybenzylidene)aniline ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M0582-25G
M0582-5G
TCIAM0582-25GEA
90.2
EUR
TCIAM0582-25G
TCIAM0582-5G
N-(4-Methoxybenzylidene)aniline ≥98.0% (by GC, titration analysis)
N-(4-Methoxybenzylidene)aniline
Formule:
C₁₄H₁₃NO |
MDL Nummer:
MFCD00025836 CAS nummer: 836-41-9 EINECS: 212-647-8 |
|
Specification Test Results
Appearance | White to Light red to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 61.0 to 67.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...