2,4-Dimethoxy-4'-hydroxybenzophenone ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

D2707-25G D2707-5G
TCIAD2707-25GEA 186 EUR
TCIAD2707-25G TCIAD2707-5G
2,4-Dimethoxy-4'-hydroxybenzophenone ≥98.0% (by titrimetric analysis)
2,4-Dimethoxy-4'-hydroxybenzophenone
Formule: C₁₅H₁₄O₄
MDL Nummer: MFCD00137850
CAS nummer: 41351-30-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5040f02015be589955ff6b870efb39179990f385&vid=5040f02015be589955ff6b870efb39179990f385&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 93.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 150.0 to 155.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR