4-Methoxybenzoic anhydride ≥97.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

M1973-5G M1973-25G
TCIAM1973-5GEA 107 EUR
TCIAM1973-5G TCIAM1973-25G
4-Methoxybenzoic anhydride ≥97.0% (by GC, titration analysis)
4-Methoxybenzoic anhydride
Formule: C₁₆H₁₄O₅
Kookpunt: 310°C
MDL Nummer: MFCD00017175
CAS nummer: 794-94-5
EINECS: 212-345-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94c0616c3eea4fff4fd1150be1102b7919395b58&vid=94c0616c3eea4fff4fd1150be1102b7919395b58&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Purity (Morpholine Method) min. 97.0 %
Melting point 96.0 to 100.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR