2-Chloronicotinoyl chloride ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
C1604-25G C1604-5G
TCIAC1604-25GEA 150 EUR
TCIAC1604-25G TCIAC1604-5G
2-Chloronicotinoyl chloride ≥98.0% (by titrimetric analysis)
2-Chloronicotinoyl chloride
Formule: C₆H₃Cl₂NO
Kookpunt: 120°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00051677
CAS nummer: 49609-84-9
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e490e8a66219e5715fb057cbe0f6f34c9b979c50&vid=e490e8a66219e5715fb057cbe0f6f34c9b979c50&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Light yellow powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Melting point 38.0 to 42.0 deg-C