3-Fluoro-4-(trifluoromethoxy)aniline
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC5548-1G PC5548-5G PC5548-10G
APOSPC5548-1GEA 33.7 EUR
APOSPC5548-1G APOSPC5548-5G APOSPC5548-10G
3-Fluoro-4-(trifluoromethoxy)aniline
3-Fluoro-4-(trifluoromethoxy)aniline
Formule: C₇H₅F₄NO
MW: 195.12 g/mol
MDL Nummer: MFCD09832383
CAS nummer: 1017779-69-9
UN: 2810
ADR: 6.1 III