Decyl chloroformate ≥95.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

C1357-25ML C1357-5ML
TCIAC1357-25MLEA 152 EUR
TCIAC1357-25ML TCIAC1357-5ML
Decyl chloroformate ≥95.0% (by titrimetric analysis)
Decyl chloroformate
Formule: C₁₁H₂₁ClO₂
Kookpunt: 163°C
MDL Nummer: MFCD00126853
CAS nummer: 55488-51-2
EINECS: 259-668-9
UN: 3277
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c731964db6f608ddd38b06cc172b01c5c56675f&vid=8c731964db6f608ddd38b06cc172b01c5c56675f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (Argentmetric Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR