Hexanophenone 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

120720250 120720050
ACRO120720250EA 76.6 EUR
ACRO120720250 ACRO120720050
Hexanophenone 98%
Hexanophenone
Formule: C₁₂H₁₆O
Kookpunt: 265,1 °C (760 mmHg)
Smeltpunt: 25…26 °C
Dichtheid: 0,95 g/cm³
Vlampunt: >112 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009512
CAS nummer: 942-92-7
EINECS: 213-394-6

Order Now

Specification Test Results

Appearance Clear light yellow Liquid after melting
Infrared spectrum Conforms
GC ≥97.5 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR