1-Phenyl-1-butanol ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P1682-25G P1682-5G
TCIAP1682-25GEA 191 EUR
TCIAP1682-25G TCIAP1682-5G
1-Phenyl-1-butanol ≥98.0% (by GC)
1-Phenyl-1-butanol
Formule: C₁₀H₁₄O
Kookpunt: 170°C
Smeltpunt: 16°C
MDL Nummer: MFCD00066173
CAS nummer: 614-14-2
EINECS: 210-368-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3c758801f181b2c58aeac547b91e3379621172d&vid=f3c758801f181b2c58aeac547b91e3379621172d&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %