Benzamidoxime
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
RF01827DA
RF01827EA
MAYBRF01827DAEA
0
EUR
MAYBRF01827DA
MAYBRF01827EA
Benzamidoxime
Benzamidoxime
Formule:
C₇H₈N₂O |
MDL Nummer:
MFCD00031485 CAS nummer: 613-92-3 EINECS: 210-361-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...