2-Isopropyl-6-methylaniline

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SB01256.1 SB01256.10 SB01256.50
MAYBSB01256.1EA 0 EUR
MAYBSB01256.1 MAYBSB01256.10 MAYBSB01256.50
2-Isopropyl-6-methylaniline
2-Isopropyl-6-methylaniline
Formule: C₁₀H₁₅N
MDL Nummer: MFCD00007746
CAS nummer: 5266-85-3
EINECS: 226-083-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR