3,4-Bis(benzyloxy)benzaldehyde ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D4079-5G
D4079-25G
TCIAD4079-5GEA
34.1
EUR
TCIAD4079-5G
TCIAD4079-25G
3,4-Bis(benzyloxy)benzaldehyde ≥98.0% (by GC)
3,4-Bis(benzyloxy)benzaldehyde
Formule:
C₂₁H₁₈O₃ |
MDL Nummer:
MFCD00004776 CAS nummer: 5447-02-9 EINECS: 226-662-2 |
|
Specification Test Results
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 90.0 to 94.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...