4-Bromo-N-phenylaniline ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B3949-5G B3949-25G
TCIAB3949-5GEA 26.4 EUR
TCIAB3949-5G TCIAB3949-25G
4-Bromo-N-phenylaniline ≥98.0% (by GC)
4-Bromo-N-phenylaniline
Formule: C₁₂H₁₀BrN
Kookpunt: 318°C
MDL Nummer: MFCD07779504
CAS nummer: 54446-36-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 85.0 to 89.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR