5-Chloro-2-methylbenzoic acid ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 5-Chloro-o-toluic acid
C2509-5G C2509-25G
TCIAC2509-5GEA 35.2 EUR
TCIAC2509-5G TCIAC2509-25G
5-Chloro-2-methylbenzoic acid ≥98.0% (by GC, titration analysis)
5-Chloro-2-methylbenzoic acid
Formule: C₈H₇ClO₂
MDL Nummer: MFCD00045842
CAS nummer: 7499-06-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2655ca79d4997562b0f2cdfb6438411dbba110e2&vid=2655ca79d4997562b0f2cdfb6438411dbba110e2&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 171.0 to 175.0 deg-C