5-Amino-2-(trifluoromethyl)pyridine ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
3-Amino-6-(trifluoromethyl)pyridine, 2-(Trifluoromethyl)pyridin-5-amine, 2-(Trifluoromethyl)-5-pyridylamine
A1816-5G
A1816-1G
TCIAA1816-5GEA
363
EUR
TCIAA1816-5G
TCIAA1816-1G
5-Amino-2-(trifluoromethyl)pyridine ≥98.0% (by GC, titration analysis)
5-Amino-2-(trifluoromethyl)pyridine
Formule:
C₆H₅F₃N₂ MW: 162.11 g/mol |
MDL Nummer:
MFCD01862656 CAS nummer: 106877-33-2 |
|
Specification Test Results
Appearance | White to Brown powder to crystalline |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 42.0 to 46.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...