Brilliant cresyl blue ALD, certified by the Biological Stain Commission, Sigma-Aldrich®
Leverancier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: CI 51010, 1,3-Diamino-7-(diethylamino)-3-methylphenoxazin-5-ium tetrachlorozincate (2:1), Brilliant cresyl blue zinc chloride double salt, Brilliant Blue C, BCB
M-Clarity™ quality level = MQ200
860867-10G 860867-25G
860867-10GEA 113 EUR
860867-10G 860867-25G
Brilliant cresyl blue ALD, certified by the Biological Stain Commission, Sigma-Aldrich®
Brilliant cresyl blue
Brilliant cresyl blue ALD has been used for the staining of reticulocytes.
The structure of Brilliant Cresyl Blue ALD contains a 2-methyl modification on the phenoxazine core, while the structure of the historic Brilliant Cresyl Blue contains a 8-methyl modification on the phenoxazine core. The biological staining application is the same for both structures.
Formule: C₃₄H₄₂Cl₄N₈O₂Zn
MW: 801.96 g/mol
MDL Nummer: MFCD00148901
CAS nummer: 81029-05-2
EINECS: 279-675-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ba87fe9d53dcd67d47b0a617a0702325dd9d979&vid=68985ea7723e1e95168fb568a6de3ba76db42f89&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Dye content ≥50%