3-Bromosalicylaldehyde ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B3922-5G B3922-1G
TCIAB3922-5GEA 162 EUR
TCIAB3922-5G TCIAB3922-1G
3-Bromosalicylaldehyde ≥98.0% (by GC, titration analysis)
3-Bromosalicylaldehyde
Formule: C₇H₅BrO₂
MDL Nummer: MFCD00016587
CAS nummer: 1829-34-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cd40aca752d2bffcc32e4e04e2f9d7c24c1110b&vid=7cd40aca752d2bffcc32e4e04e2f9d7c24c1110b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 54.0 to 58.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR