4-Fluoro-2-methylbenzaldehyde ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 4-Fluoro-2-tolualdehyde, 4-Fluoro-o-tolualdehyde, 4-Fluoro-o-tolylaldehyde

F0793-1G F0793-5G
TCIAF0793-1GEA 17.6 EUR
TCIAF0793-1G TCIAF0793-5G
4-Fluoro-2-methylbenzaldehyde ≥95.0% (by GC)
4-Fluoro-2-methylbenzaldehyde
Formule: C₈H₇FO
Kookpunt: 62°C
Vlampunt: 82°C
MDL Nummer: MFCD02261764
CAS nummer: 63082-45-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab0bccb8f21070cec6c33345e8dbd40017060095&vid=ab0bccb8f21070cec6c33345e8dbd40017060095&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR