Trimethylamine ∼28% (w/w) ∼4.3 mol/L in water
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
T0464-25ML T0464-500ML
TCIAT0464-25MLEA 18.7 EUR
TCIAT0464-25ML TCIAT0464-500ML
Trimethylamine ∼28% (w/w) ∼4.3 mol/L in water
Trimethylamine
Formule: C₃H₉N
Kookpunt: 30 °C (1013 hPa)
Smeltpunt: –3 °C
Dichtheid: 0,85 to 0.9 g/cm³ (20 °C)
Vlampunt: 3°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00008327
CAS nummer: 75-50-3
EINECS: 200-875-0
UN: 1297
ADR: 3,II
Merck Index: 14,09710
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5591e14b9e32e2d34918ad382bcd7050f23cb5f&vid=f5b8f4a12cb7334a45c577c9c38879a250788549&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless clear liquid
Concentration (Neutralization titration) 28.0 to 31.0 w/w%