Thiobenzoylmercaptoacetic acid ≥97.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

T2401-25G T2401-5G
TCIAT2401-25GEA 268 EUR
TCIAT2401-25G TCIAT2401-5G
Thiobenzoylmercaptoacetic acid ≥97.0% (by GC, titration analysis)
Thiobenzoylmercaptoacetic acid
Formule: C₉H₈O₂S₂
MDL Nummer: MFCD00004927
CAS nummer: 942-91-6
EINECS: 213-393-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81af652ef90ee9f5bffe81014160cbeca42ce0ed&vid=81af652ef90ee9f5bffe81014160cbeca42ce0ed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Orange to Brown to Dark red powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 126.0 to 130.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR