Photographic trays
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0654 0655 0658
SEMA0654EA 153 EUR
SEMA0654 SEMA0655 SEMA0658
Photographic trays
Reservoirs/Schalen/Potten Laboratoriumbakken/-schalen
Trays made of PVC, inside white and outside grey in colour, with floor grooves.
  • Stable and cost effective
  • Resistant to acids
  • Temperature resistance up to 60 °C