Dosing bottles, LDPE
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1208 1211 1210
215-0280EA 15 EUR
215-0280 215-0291 SEMA1210
Dosing bottles, LDPE
Flessen Druppelflessen
LDPE, translucent dosing bottles attached white PP cap with pouring spout.
  • Good chemical resistance
  • Good temperature resistance
Ideal for fast and clean dosing and sampling of the desired liquid quantity.
Opgelet: Not suitable for shipping liquids.