Bottles, narrow neck, round or square, HDPE, with screw caps
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4585 4291
SEMA4585EA 153 EUR
SEMA4585 SEMA4291
Bottles, narrow neck, round or square, HDPE, with screw caps
Flessen Kunststof flessen
HDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
  • Un approved 3H1/X250 for fluids
With tamper evident screw cap.