Bottles, wide neck, HDPE, round, with screw cap
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3030 3031
SEMA3030EA 8.9 EUR
SEMA3030 SEMA3031
Bottles, wide neck, HDPE, round, with screw cap
Flessen Kunststof flessen
HDPE, amber.
  • Good chemical resistance
  • Good temperature resistance
  • Tight closure
Amber coloured to protect light sensitive substances.