Container with snap-on lid, Azlon®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JWH1000P JWH2500P JWH0500P JWH0125P JWH0250P JWH2000P JWH5000P
AZLOJWH1000PEA 308 EUR
AZLOJWH1000P AZLOJWH2500P AZLOJWH0500P AZLOJWH0125P AZLOJWH0250P AZLOJWH2000P AZLOJWH5000P
Container with snap-on lid, Azlon®
Monsternamepotjes Containers voor algemeen gebruik
PP, translucent.
  • Good chemical resistance
  • Suitable for food contact applications
  • Very good temperature resistance
These containers are lightweight tapered with tight fitting lids. Ideal for storage of samples, powders and many more.
Order Now
Specificaties
Environmentally Preferable