Non return valves, Azlon®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
NWH902P NWH906P
AZLONWH902PEA 70.1 EUR
AZLONWH902P AZLONWH906P
Non return valves, Azlon®
Ventielen Terugslagventielen
Designed for use in vacuum lines and fluid handling systems.
  • Useful for preventing back pressure in filter flasks
  • Printed arrow indicates the direction of flow
  • Rigid with stepped tapered arms to give firm grip on tubing