Absorbent pillows, NovaSpill™
Leverancier: MICRONOVA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
NS6-28 NS6-28IR
121-0134EA 45.4 EUR
121-0134 121-0135
Absorbent pillows, NovaSpill™
Absorptiematerialen en bescherming tegen morsen
An absorbent sock/pillow (711 mm) which consists of polyester strips contained in a polyester sleeve.
  • Low linting with good chemical resistance
  • Easy to position for spill containment
  • Can be autoclaved if required
They are designed for cleanroom use.