Flathead mop covers, SnapMop™
Leverancier: MICRONOVA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MZSM620-14 MZSM6AA-14 MZSM56-9 MZSM56-9IR MZSM620-14IR MZSM620-18 MZSM620-18IR MZSM620-9 MZSM6AA-18 MZSM6AA-18IR MZSM6AA-9 MZSM6AA-9IR MZSM6AW-14 MZSM6AW-18 MZSM11-14NS
129-0974EA 30.6 EUR
129-0974 129-0975 141-1021 141-1022 141-1023 141-1024 141-1025 141-1026 141-1029 141-1030 141-1031 141-1032 MICRMZSM6AW-14 MICRMZSM6AW-18 MICRMZSM11-14NS
Flathead mop covers, SnapMop™
Moppen
A range of lightweight, snap-on flat head mop covers with urethane foam interiors, designed for effective cleaning and sanitising in the cleanroom. A variety of fabric options provide excellent absorbency, effective cleaning, and disinfectant dispersal for specialised cleaning.
  • Easy to change covers fit securely over mop frame
  • Urethane foam interiors fully encased in a choice of fabrics
  • NovaPoly: 100% smooth knit polyester provides excellent disinfectant dispersal
  • PolySorb: 100% textured polyester for removing residues and dispersing disinfectant
  • MegaTex: 100% non-woven polyamide is absorbent with a mildly abrasive surface
  • PolyMesh: 100% polyester with a layer of mesh provides a mild scrubbing surface
  • 228,6 mm covers are excellent for pass-throughs and smaller spaces
  • 355,6 and 457,2 mm covers are ideal for floors, walls, and ceilings
Covers are constructed with a thin urethane foam interior completely encased in polyester, ensuring no exposed foam and resulting in less weight and faster drying. Available in 100% polyester or with 'cleaning bands' in microfibre for additional cleaning power or MegaTex® for cleaning rough/textured surfaces or removing heavy deposits. Can be laundered, re-used and able to withstand alcohol, strong disinfectants and steam sterilisation. Also available gamma irradiated.
Stainless steel snap fasteners connection.