Isolator Cleaning Tools
Leverancier: MICRONOVA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
QDAH-13 QDIT-09 ITC6-9IR QPIT-09 QDAH-19 ITC11-9NSIR STAH-12WH ITC620-911IR QDAH-08 ITCAW-9 ITCAA-9 ITC6-911NSIR ITC11-9NS ITC6-9 ITC6-9NSIR ITC11-9NSBB
129-0622EA 298 EUR
129-0622 129-0625 129-0611 216-1490 129-0623 MICRITC11-9NSIR 141-0897 MICRITC620-911IR MICRQDAH-08 MICRITCAW-9 MICRITCAA-9 MICRITC6-911NSIR MICRITC11-9NS MICRITC6-9 MICRITC6-9NSIR MICRITC11-9NSBB
Isolator Cleaning Tools
Moppen
The Isolator Tool™ adapter with a 360° swivel works with a range of mop covers to efficiently clean and disinfect isolators, hoods, RABS, and small equipment in clean and aseptic areas.
  • Low-profile design and angled head adapter
  • Available with stainless steel or plastic plates
  • PolySorb™: 100% textured polyester for additional cleaning as well as sanitisation
  • NovaPoly™: 100% smooth knit polyester provides excellent disinfectant dispersal
  • MegaTex®: Polyamide, slightly abrasive yet absorbent
  • Microfibre: 80% Polyester/20% Nylon for maximum absorbency and residue removal
  • Optional urethane foam layer to hold disinfectant and facilitate wringing
  • Compatible with alcohol and strong disinfectants
  • Launderable for reuse
  • Autoclavable
Designed for use with the SSU series Universal Handles or STA series threaded handles. Use modular and extendable handles to fit through small passes and allow extended reach.
Handles: Straight handles are available in electropolished stainless steel or plastic. Accordion handles are designed to deal with the unique space limitations in isolators and containment hoods. Available in 330 and 483 mm sizes, each section features a delrin hinge and quick disconnect to allow for attachment of additional sections or isolator tool itself.
Bestelinformatie: Fabric covers, adapters, and handles are ordered separately. The all plastic Isolator Tool™ and slip covers are available gamma irradiated, contact your Avantor Sales Representative for more information.