Capillary columns, TRACE™ TR-5MS
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
260F396P 260F578P 260F336P 260F309P 260F035P 260F047P 260F130P 260F142P 260F143P 260F020P 260F223P 260F224P 260F225P 260F285P 260F296P 260F297P 260F298P 260F308P 260F154P
555-9496EA 920 EUR
555-9496 555-9497 555-9494 555-9493 555-9464 555-9463 555-9462 555-9458 555-9457 555-9466 555-9452 555-9451 555-9450 555-9447 555-9446 555-9445 555-9444 555-9474 555-9455
Capillary columns, TRACE™ TR-5MS
Kolommen voor chromatografie
TRACE™ TR-5MS GC columns are ideal for hydrocarbons, solvents, pesticides, herbicides, phenols, amines.
  • Non-polar phase, 5% phenyl polysilphenylene-siloxane
  • Low bleed and high stability
  • High signal-to-noise for increased sensitivity
  • High robustness to oxygen and water contamination