Capillary columns, TRACE™ TR-5
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
260E242P 260E113P 260E130P 260E131P 260E142P 260E143P 260E154P 260E155P 260E225P 260E297P 260E298P 260E336P 260E470P 260E224P
555-9291EA 1540 EUR
555-9291 555-9301 555-9300 555-9299 555-9298 555-9297 555-9296 555-9295 555-9292 555-9288 555-9287 555-9470 555-9469 555-9293
Capillary columns, TRACE™ TR-5
Kolommen voor chromatografie
Excellent starting columns for method development, capable of performing most required separations.
  • Non-polar phase, 5% phenyl methyl polysiloxane
  • High operating temperature and extremely low bleed
  • Widely used in a variety of applications