Safety blood lancets
Leverancier: Sarstedt
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SARS85.1015EA 124 EUR
SARS85.1015 SARS85.1017 SARS85.1016 SARS85.1019 SARS85.1018
Safety blood lancets
Bloedlancetten
Safety blood lancets for safe capillary blood collection and increased patient comfort.
  • User and patient safety
  • Increased patient comfort
  • Designed for targeted blood flow
It reduces the risk of needle injury and cross contamination because the needle point or blade is concealed both before and after use. The sterile device is for single use only and can be safely discarded.
Bestelinformatie: A range of penetration depths and widths are available to accommodate various volume requirements and skin types, ensuring optimum blood flow.