Resin, PL-Wang, StratoSpheres™
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PL1463-6749 PL1463-6769 PL1463-6799 PL1463-6799HL
VARIPL1463-6749EA 0 EUR
VARIPL1463-6749 VARIPL1463-6769 VARIPL1463-6799 VARIPL1463-6799HL
Resin, PL-Wang, StratoSpheres™
Sorbenten voor chromatografie Sorbenten voor SPE
PL-Wang is a 4-alkoxybenzylalcohol functionalised polystyrene, prepared from copolymerised PL-CMS, for solid phase peptide synthesis using Fmoc protection strategies and is cleaved using ~ 95% TFA. Particularly useful for solid phase synthesis of small molecules which have a carboxylic acid functional group.
  • Amino acids and carboxylic acids are attached to this resin through esterification
  • Care should be taken during the activation procedure to minimise any risk of racemisation
Acidic alcohols, particularly phenols, may also be attached to PL-Wang, these resins have also been converted to carbamate functionalised materials in order to prepare substituted amines.
Order Now
Specificaties
Environmentally Preferable