Capillary columns, TraceGOLD™ TG-1701MS
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
26090-3080 26090-2240 26090-2350 26090-2960 26090-2980 26090-3090 26090-3960 26090-2970 26090-2230 26090-0630 26090-1310 26090-1420 26090-1430 26090-1440 26090-1540 26090-1550
555-8572EA 1440 EUR
555-8572 555-8513 555-8519 555-8525 555-8573 555-8571 555-8564 555-8526 555-8512 555-8536 555-8533 555-8531 555-8530 555-8529 555-8528 555-8527
Capillary columns, TraceGOLD™ TG-1701MS
Kolommen voor chromatografie
These columns feature a mix of cyano and phenyl groups for increased polarity and a different elution order relative to less polar columns.
  • Mid-polarity phase, 14% cyanopropylphenyl, 86% dimethyl polysiloxane
  • Fully characterised for long-term reproducibility, column-to-column consistency and low bleed
  • Optimal for confirmation analysis
  • Equivalent to USP G46 phase