Capillary columns, TraceGOLD™ TG-17MS
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
26089-2960 26089-2970 26089-2980 26089-3360 26089-2230 26089-2240 26089-1300 26089-1420 26089-1430 26089-1440
555-8542EA 789 EUR
555-8542 555-8541 555-8540 555-8538 555-8610 555-8609 555-8620 555-8617 555-8616 555-8615
Capillary columns, TraceGOLD™ TG-17MS
Kolommen voor chromatografie
These columns are particularly suited to GCMS applications that require more polarity than a 5% phenyl phase.
  • Mid-polarity phase of 50% diphenyl or 50% dimethyl polysiloxane
  • Ideal for confirmational analysis
  • Excellent inertness for active compounds such as pesticides
  • Very low bleed ideal for analysis by GCMS