Eenkanaalspipetten, mechanisch, variabel volume, PIPETMAN® L
Leverancier: GILSON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FA10008 FA10007 FA10006M FA10003P FA10005P FA10006P FA10003M FA10005M FA10004P FA10004M FA10001M FA10001P FA10002M FA10002P F167350 F167550 F167370 FA10001MC FA10001MCG FA10001PC FA10001PCG FA10002MC FA10002MCG FA10002PC FA10002PCG FA10003MC FA10003MCG FA10003PC FA10003PCG FA10004MC FA10004MCG FA10004PC FA10004PCG FA10005MC FA10005MCG FA10005PC FA10005PCG FA10006MC FA10006MCG FA10006PC FA10006PCG FA10007C FA10007CG FA10008C FA10008CG
613-5813EA 342 EUR
613-5813 613-5812 613-5805 613-5808 613-5810 613-5811 613-5802 613-5804 613-5809 613-5803 613-5800 613-5806 613-5801 613-5807 613-5890 613-5891 613-5892 GILSFA10001MC GILSFA10001MCG GILSFA10001PC GILSFA10001PCG GILSFA10002MC GILSFA10002MCG GILSFA10002PC GILSFA10002PCG GILSFA10003MC GILSFA10003MCG GILSFA10003PC GILSFA10003PCG GILSFA10004MC GILSFA10004MCG GILSFA10004PC GILSFA10004PCG GILSFA10005MC GILSFA10005MCG GILSFA10005PC GILSFA10005PCG GILSFA10006MC GILSFA10006MCG GILSFA10006PC GILSFA10006PCG GILSFA10007C GILSFA10007CG GILSFA10008C GILSFA10008CG
Eenkanaalspipetten, mechanisch, variabel volume, PIPETMAN® L
Pipetten Micropipetten
Designed for performance and comfort, PIPETMAN® L comes in a comprehensive range of models, adapted to your needs in the lab. PIPETMAN L air-displacement pipettes cover a full volume range. Comfortable, precise, accurate, and reliable, use PIPETMAN L pipettes confidently in your applications.
  • Convenient use
  • Secure pipetting
  • GLP compliance
Lightweight and balanced, PIPETMAN L combines an ergonomic design with minimized pipetting forces, giving you the ability to pipette longer without fatigue. Right-handed or left-handed? No problem. Adjust the location of the tip ejector to the most comfortable position for you.
PIPETMAN L single channel models are equipped with a stainless-steel tip ejector. Should your application require to pipette corrosive liquids, plastic tip ejectors are available as accessories. P5000L and P10mLL models are equipped with a plastic tip ejector to avoid direct contact with the tip and provide better compatibility with chemicals.
Reduce pipetting errors and avoid accidental volume changes during pipetting cycles with a true volume-locking mechanism for improved accuracy and precision. PIPETMAN L models are all fully autoclavable without disassembly for maximum convenience and protection from cross-contamination.
PIPETMAN L models feature a DataMatrix 2D code for easy traceability. Use a 2D code scanner to transfer the code automatically into any traceability or asset management system. Also, the pipette serial number is engraved on the body of the pipette for unique identification.
Leveringsinformatie: De Microvolume startkit bevat de drie pipetten P2L (0,2 - 2 µl), P10L (1 - 10 µl) en P100L (10 - 100 µl), twee rekken van DL10 en D200 DIAMOND tips, drie SINGLE™ pipethouders, poster 'inspectie in twee minuten', Gilson pipetteerhandleiding en drie kunststof ejectors.
De Pipetman® L startkit bevat de drie pipetten P20L (2 - 20 µl), P200L (20 - 200 µl) en P1000L (100 - 1000 µl), twee rekken van D200 en D1000 DIAMOND tips, drie SINGLE™ pipethouders, poster 'inspectie in twee minuten', Gilson pipetteerhandleiding en drie kunststof ejectors.
De Pipetman® L startkit '4-pipette' bevat de vier pipetten P2L (0,2 - 2 µl), P20L (2 - 20 µl), P200L (20 - 200 µl) en P1000L (100 - 1000 µl) met roestvrijstalen tipejector, drie rekken van DL10, D200 en D1000 DIAMOND tips, vier SINGLE™ pipethouders, poster 'inspectie in twee minuten' en Gilson pipetteerhandleiding.