Reagent reservoirs
Leverancier: GILSON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F267660 F267670
GILSF267660EA 155.4 EUR
GILSF267660 GILSF267670
Reagent reservoirs
Reagensreservoirs
These reagent reservoirs are ideal for dispensing reagents and solutions using a multichannel pipette. The disposable, white PS reservoirs are available in 25-mL and 50-mL sizes.
  • Wide stable bases, angled sides, and V-shaped bottoms allow for efficient and nearly complete removal of solutions
  • Reservoirs provide spouts on all corners to simplify emptying and reduce spillage when removing liquids
  • Middle volume graduations on the inside walls for quick measurement of liquids within the trough