Laboratory bottles, narrow neck, with or without screw cap
Leverancier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
632414315966 632414321251 632414321501 632414315500 632414315940 632414315950 632414315956 632414315100 632414315250
215-1647EA 454 EUR
215-1647 KAVA632414321251 KAVA632414321501 KAVA632414315500 KAVA632414315940 KAVA632414315950 KAVA632414315956 KAVA632414315100 KAVA632414315250
Laboratory bottles, narrow neck, with or without screw cap
Flessen Glazen flessen
Borosilicate glass 3,3, GL 45 threaded, with or without scew cap.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Round, with graduation, DIN GL 45 thread, ETFE pouring ring and autoclavable red PBT screw cap (temperature resistance up to 180 °C) with PTFE coated silicone gasket.
Certificaten: In compliance with DIN ISO 4796 - 1 standards.