Detergent, alkaline Cleaning-In-Place detergents, Amethyst and Topaz
Leverancier: TRISTEL SOLUTIONS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CRY404 CRY405 CRY403 CRY404/1L CRY401/1L
TRSTCRY404EA 0 EUR
TRSTCRY404 TRSTCRY405 TRSTCRY403 TRSTCRY404/1L TRSTCRY401/1L
Detergent, alkaline Cleaning-In-Place detergents, Amethyst and Topaz
Reinigingsmiddelen Detergenten en oppervlaktereinigers
A non foaming, medium duty caustic based, CIP detergent for the removal of organic soils, including fats and oils.
  • Medium duty detergent; for use on organic and inorganic soils
  • Suitable for use in CIP and spraywash enclosed systems
  • Dilution for use: amethyst - 1 to 5% v/v; topaz - 0,5 to 5% v/v
Areas of use include equipment such as CIP and spray wash enclosed systems. Amethyst and Topaz are not soft metal safe, but are suitable for use on stainless steel.
Crystel offer highly effective Cleaning-In-Place (CIP) detergents for automated processes within manufacturing sites. The product range is designed to remove a variety of residues that can be encountered. Residue test methods are available for all CIP detergents offering you peace of mind that there will be reduced risk of contamination of your products.
All detergents are supported by comprehensive technical files which provide all the documentation for validation, including residue test methods.
Leveringsinformatie: Supplied with its own certificate of analysis. A detailed technical file (including: product specification, manufacturing process, shelf life, microbiological and analytical data) is available on request.
Verpakking: Available in 20, 200 and 1000 litre pack sizes.