Maatcilinders, hoog model, klasse A, borosilicaatglas
Leverancier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
632432110718 632432110819 632432110923 632432111125 632432111130 632432111238 632432111343 632432111444 632432111546
KAVA632432110718EA 292 EUR
KAVA632432110718 KAVA632432110819 KAVA632432110923 KAVA632432111125 KAVA632432111130 KAVA632432111238 KAVA632432111343 KAVA632432111444 KAVA632432111546
Maatcilinders, hoog model, klasse A, borosilicaatglas
Cilinders Maatcilinders
Borosilicate glass 3,3, tall form, class A.
  • Hexagonal glass base, with spout
  • Blue graduations
Certificaten: ISO 4788, DIN 12680