Microtiter tubes system
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3487 3492 3496 3425-11 3426-11
MLBP3487EA 66.8 EUR
MLBP3487 MLBP3492 MLBP3496 MLBP3425-11 MLBP3426-11
Microtiter tubes system
Buisjes Microbuisjes
Tubes made of PP, natual, caps made of PE, natural.
  • Robotic compatible rack can easily integrated into automated liquid handling workstations
  • Polyethylene plug strip seals the microtiter tubes tightly for storage
  • Eight plug strips can be easily cut apart for individual samples
Microtiter tubes have additional volume capacity for sample dilution and are ideal for HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control and blood bank sample freezer storage. Frosted writing areas on each tube and an alphanumeric rack simplify organization and identification of specific samples.
Certificaten: Non sterile: certified DNA, RNase/DNase, pyrogen and endotoxin-free.
Sterile: certified DNA, RNase/DNase, pyrogen, endotoxin, PCR inhibitor, and bioburden-free.