2-Methoxyestradiol, Calbiochem®
Leverancier: Merck Millipore Calbiochem
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: (8R,9S,13S,14S,17S)-2-Methoxy-13-methyl-7,8,9,11,12,13,14,15,16,17-decahydro-6H-cyclopenta[a]phenanthrene-3,17-diol
454180-10 454180-50
454180-10EA 116 EUR
454180-10 454180-50
2-Methoxyestradiol, Calbiochem®
2-Methoxyestradiol
Formule: C₁₉H₂₆O₃
MW: 302.408 g/mol
MDL Nummer: MFCD00010489
CAS nummer: 362-07-2