Arachidonic acid, Millipore®
Leverancier: Merck Millipore Calbiochem
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
181198-1 181198-100
181198-1EA 400 EUR
181198-1 181198-100
Arachidonic acid, Millipore®
Arachidonic acid
Formule: C₂₀H₃₂O₂
MDL Nummer: MFCD00004417
CAS nummer: 506-32-1
EINECS: 208-033-4