4-Methoxyisophthalaldehyde

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

RH01260.250 RH01260.1
MAYBRH01260.250EA 0 EUR
MAYBRH01260.250 MAYBRH01260.1
4-Methoxyisophthalaldehyde
4-Methoxyisophthalaldehyde
Formule: C₉H₈O₃
CAS nummer: 25445-35-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR