5-Allyl-3-methoxysalicylaldehyde ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 5-Allyl-2-hydroxy-3-methoxybenzaldehyde

A2104-1G A2104-5G
TCIAA2104-1GEA 41.8 EUR
TCIAA2104-1G TCIAA2104-5G
5-Allyl-3-methoxysalicylaldehyde ≥98.0% (by GC, titration analysis)
5-Allyl-3-methoxysalicylaldehyde
Formule: C₁₁H₁₂O₃
Kookpunt: 167°C
MDL Nummer: MFCD06408022
CAS nummer: 22934-51-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b1f0fb8c7cdd47e77fbd22db58f22d2770010dd&vid=0b1f0fb8c7cdd47e77fbd22db58f22d2770010dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point gt. 46.0 to 50.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR