(tert-Butylamino)triphenylphosphonium bromide
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JFD02770.250
JFD02770.1
MAYBJFD02770.250EA
0
EUR
MAYBJFD02770.250
MAYBJFD02770.1
(tert-Butylamino)triphenylphosphonium bromide
(tert-Butylamino)triphenylphosphonium bromide
Formule:
C₂₂H₂₅BrNP |
CAS nummer:
799-51-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...