Amino(triphenyl)phosphonium chloride
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BTB11223.250
BTB11223.1
MAYBBTB11223.250EA
0
EUR
MAYBBTB11223.250
MAYBBTB11223.1
Amino(triphenyl)phosphonium chloride
Amino(triphenyl)phosphonium chloride
Formule:
C₁₈H₁₇ClNP |
MDL Nummer:
MFCD00032110 CAS nummer: 15729-44-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...