Ethyl dibromofluoroacetate ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
D2530-25G
D2530-5G
TCIAD2530-25GEA
262
EUR
TCIAD2530-25G
TCIAD2530-5G
Ethyl dibromofluoroacetate ≥97.0% (by GC)
Ethyl dibromofluoroacetate
Formule:
C₄H₅Br₂FO₂ Kookpunt: 173°C |
MDL Nummer:
MFCD00042068 CAS nummer: 565-53-7 UN: 2927 ADR: 6.1,II |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...