4-Pentenyl acetate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

A1345-25ML A1345-500ML
TCIAA1345-25MLEA 64.9 EUR
TCIAA1345-25ML TCIAA1345-500ML
4-Pentenyl acetate ≥98.0% (by GC)
4-Pentenyl acetate
Formule: C₇H₁₂O₂
Kookpunt: 151°C
Vlampunt: 45°C
MDL Nummer: MFCD00008723
CAS nummer: 1576-85-8
EINECS: 216-413-6
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=476ec96a2f7474e00978cccee00c7ede72f04f15&vid=476ec96a2f7474e00978cccee00c7ede72f04f15&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR