Tetrabutylphosphonium hexafluorophosphate ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

T2007-25G T2007-5G
TCIAT2007-25GEA 131 EUR
TCIAT2007-25G TCIAT2007-5G
Tetrabutylphosphonium hexafluorophosphate ≥98.0% (by titrimetric analysis)
Tetrabutylphosphonium hexafluorophosphate
Formule: C₁₆H₃₆F₆P₂
MDL Nummer: MFCD01863114
CAS nummer: 111928-21-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65109cb78eaf8e34e80f34a358f490e84ebd2bde&vid=65109cb78eaf8e34e80f34a358f490e84ebd2bde&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %
Melting point 213.0 to 221.0 deg-C

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR